Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003571.1 | NP_001003571.1 | 1 - 2 | 467 | 89 | |
GenBank mRNA | BC077119.1 | - | 1 - 2 | 506 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |