Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003840.1 | NP_001003840.1 | 3 - 4 | 403 | 101 | |
GenBank mRNA | AY648829.1 | - | 3 - 4 | 403 | 101 | |
BC153551.1 | - | 3 - 4 | 373 | 101 | ||
BC162284.1 | - | 3 - 4 | 401 | 101 | ||
BC162298.1 | - | 3 - 4 | 401 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |