Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005981.1 | NP_001005981.1 | 5 - 6 | 718 | 89 | |
XM_005170550.4 | XP_005170607.1 | 5 - 6 | 820 | 89 | ||
GenBank mRNA | BC083429.1 | - | 5 - 6 | 718 | 89 | |
BC163998.1 | - | 5 - 6 | 701 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |