Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006024.1 | NP_001006024.1 | 4 - 5 | 569 | 69 | |
GenBank mRNA | BC083397.1 | - | 4 - 5 | 569 | 69 | |
BC164011.1 | - | 4 - 5 | 543 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |