Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli1rs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008780.1 | NP_001008780.1 | 5 - 6 | 775 | 61 | |
XM_005158200.3 | XP_005158257.1 | 5 - 6 | 61 | |||
XM_005158202.4 | XP_005158259.1 | 5 - 6 | 61 | |||
XM_009292528.3 | XP_009290803.1 | 5 - 6 | 61 | |||
XM_009292530.3 | XP_009290805.1 | 5 - 6 | 61 | |||
GenBank mRNA | AY839950.1 | - | 5 - 6 | 775 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |