Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pomgnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012384.2 | NP_001012384.2 | 2 - 3 | 493 | 60 | |
GenBank mRNA | AJ868539.1 | - | 2 - 3 | 493 | 60 | |
BC095667.1 | - | 2 - 3 | 412 | 60 | ||
DT073564.1 | - | 2 - 3 | 432 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |