Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: azin1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC092747.1 | - | 5 - 6 | 733 | 109 | |
BC164997.1 | - | 3 - 4 | 486 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |