Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017644.1 | NP_001017644.1 | 10 - 11 | 1014 | 73 | |
XM_009304977.3 | XP_009303252.1 | 10 - 11 | 1199 | 73 | ||
XM_017357673.2 | XP_017213162.1 | 10 - 11 | 1010 | 73 | ||
GenBank mRNA | BC092805.1 | - | 10 - 11 | 1014 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |