Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017644.1 | NP_001017644.1 | 11 - 12 | 1133 | 98 | |
XM_009304977.3 | XP_009303252.1 | 11 - 12 | 1318 | 98 | ||
XM_017357673.2 | XP_017213162.1 | 11 - 12 | 1129 | 98 | ||
GenBank mRNA | BC092805.1 | - | 11 - 12 | 1133 | 98 | |
BC154162.1 | - | 11 - 12 | 1323 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |