Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017644.1 | NP_001017644.1 | 12 - 13 | 1258 | 74 | |
XM_009304977.3 | XP_009303252.1 | 12 - 13 | 1443 | 74 | ||
XM_017357673.2 | XP_017213162.1 | 12 - 13 | 1254 | 74 | ||
GenBank mRNA | BC092805.1 | - | 12 - 13 | 1258 | 74 | |
BC154162.1 | - | 12 - 13 | 1448 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |