Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017644.1 | NP_001017644.1 | 3 - 4 | 315 | 80 | |
XM_009304977.3 | XP_009303252.1 | 3 - 4 | 500 | 80 | ||
XM_017357673.2 | XP_017213162.1 | 3 - 4 | 311 | 80 | ||
GenBank mRNA | BC092805.1 | - | 3 - 4 | 315 | 80 | |
BC154162.1 | - | 3 - 4 | 505 | 80 |
Gene Symbol: LOC100536170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021470984.1 | XP_021326659.1 | 2 - 3 | 420 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |