Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpysl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020517.2 | NP_001018353.1 | 8 - 9 | 969 | 70 | |
XM_005155631.4 | XP_005155688.1 | 9 - 10 | 1325 | 70 | ||
GenBank mRNA | AY987375.1 | - | 8 - 9 | 960 | 70 | |
AY997023.1 | - | 8 - 9 | 967 | 70 | ||
BC129137.1 | - | 8 - 9 | 969 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |