Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aamdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020764.1 | NP_001018600.1 | 2 - 3 | 198 | 88 | |
GenBank mRNA | BC095700.1 | - | 2 - 3 | 198 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |