Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aamdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020764.1 | NP_001018600.1 | 3 - 4 | 296 | 115 | |
GenBank mRNA | BC095700.1 | - | 3 - 4 | 296 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |