Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030208.1 | NP_001025379.1 | 9 - 10 | 1193 | 84 | |
GenBank mRNA | BC097128.1 | - | 9 - 10 | 1193 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |