Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nedd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034186.1 | NP_001029358.1 | 23 - 24 | 2232 | 107 | |
GenBank mRNA | AY995154.1 | - | 23 - 24 | 2232 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |