Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slit1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034968.1 | NP_001030140.1 | 33 - 34 | 3818 | 64 | |
XM_009307022.3 | XP_009305297.1 | 30 - 31 | 3537 | 64 | ||
GenBank mRNA | BC163538.1 | - | 33 - 34 | 3710 | 64 | |
DQ177286.1 | - | 33 - 34 | 3818 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |