Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slit1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034968.1 | NP_001030140.1 | 35 - 36 | 4264 | 110 | |
XM_009307022.3 | XP_009305297.1 | 32 - 33 | 3983 | 110 | ||
GenBank mRNA | BC163538.1 | - | 35 - 36 | 4156 | 110 | |
DQ177286.1 | - | 35 - 36 | 4264 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |