Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040307.1 | NP_001035397.1 | 4 - 5 | 773 | 98 | |
XM_021475840.1 | XP_021331515.1 | 4 - 5 | 852 | 98 | ||
GenBank mRNA | BC115288.1 | - | 4 - 5 | 809 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |