Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn12aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044922.1 | NP_001038387.1 | 19 - 20 | 3611 | 76 | |
XM_009298901.3 | XP_009297176.1 | 20 - 21 | 76 | |||
GenBank mRNA | DQ149507.1 | - | 19 - 20 | 3611 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons |