Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045564.2 | NP_001039029.2 | 3 - 4 | 354 | 100 | |
XM_021468901.1 | XP_021324576.1 | 3 - 4 | 352 | 100 | ||
GenBank mRNA | AF155581.1 | - | 3 - 4 | 252 | 100 | |
BC092728.1 | - | 3 - 4 | 264 | 100 | ||
BC135050.1 | - | 3 - 4 | 266 | 100 | ||
BC163781.1 | - | 3 - 4 | 315 | 100 | ||
BC164627.1 | - | 3 - 4 | 292 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |