Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zranb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077768.1 | NP_001071236.1 | 8 - 9 | 1943 | 78 | |
XM_005158595.4 | XP_005158652.1 | 9 - 10 | 2827 | 78 | ||
GenBank mRNA | BC125972.1 | - | 8 - 9 | 1943 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |