Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adpgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079965.2 | NP_001073434.1 | 6 - 7 | 997 | 73 | |
XM_005162956.4 | XP_005163013.1 | 7 - 8 | 1136 | 73 | ||
XM_005162957.4 | XP_005163014.1 | 6 - 7 | 1213 | 73 | ||
GenBank mRNA | BC125924.1 | - | 6 - 7 | 931 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |