Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mylipb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080022.2 | NP_001073491.1 | 6 - 7 | 1252 | 100 | |
GenBank mRNA | BC124450.1 | - | 6 - 7 | 1253 | 100 | |
BC165740.1 | - | 6 - 7 | 1269 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |