Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipmkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080064.1 | NP_001073533.1 | 4 - 5 | 679 | 75 | |
GenBank mRNA | BC128841.1 | - | 4 - 5 | 679 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |