Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipmkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080064.1 | NP_001073533.1 | 5 - 6 | 758 | 77 | |
GenBank mRNA | BC128841.1 | - | 5 - 6 | 758 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |