Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080184.1 | NP_001073653.1 | 4 - 5 | 595 | 64 | |
XM_021469820.1 | XP_021325495.1 | 3 - 4 | 846 | 64 | ||
GenBank mRNA | BC067342.1 | - | 4 - 5 | 564 | 64 | |
BC129317.1 | - | 4 - 5 | 595 | 64 | ||
BC164295.1 | - | 2 - 3 | 405 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |