Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2ad6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082936.1 | NP_001076405.1 | 5 - 6 | 827 | 86 | |
XM_021468529.1 | XP_021324204.1 | 2 - 3 | 293 | 86 | ||
GenBank mRNA | BC162823.1 | - | 5 - 6 | 927 | 86 | |
BC162824.1 | - | 5 - 6 | 927 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |