Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sobpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098618.1 | NP_001092088.1 | 3 - 4 | 300 | 78 | |
GenBank mRNA | DQ655703.1 | - | 3 - 4 | 300 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |