Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vsig10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100094.2 | NP_001093564.2 | 7 - 8 | 1739 | 78 | |
GenBank mRNA | CU638770.1 | - | 7 - 8 | 1739 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |