Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131233.1 | NP_571308.1 | 2 - 3 | 594 | 85 | |
XM_009305781.3 | XP_009304056.1 | 2 - 3 | 662 | 85 | ||
XM_009305782.3 | XP_009304057.1 | 2 - 3 | 799 | 85 | ||
GenBank mRNA | BC053131.1 | - | 2 - 3 | 594 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |