Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131234.2 | NP_571309.2 | 2 - 3 | 304 | 112 | |
GenBank mRNA | BC124483.1 | - | 2 - 3 | 232 | 112 | |
BC164788.1 | - | 1 - 2 | 240 | 112 | ||
U18311.1 | - | 2 - 3 | 306 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |