Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131279.2 | NP_571354.1 | 1 - 2 | 723 | 76 | |
GenBank mRNA | BC092925.1 | - | 1 - 2 | 723 | 76 | |
BC165920.1 | - | 1 - 2 | 384 | 76 | ||
D32214.1 | - | 1 - 2 | 384 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |