Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: notch1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131302.2 | NP_571377.2 | 23 - 24 | 3970 | 76 | |
XM_005168649.4 | XP_005168706.1 | 23 - 24 | 4118 | 76 | ||
GenBank mRNA | AY805407.1 | - | 23 - 24 | 3970 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |