Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ved | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183074.1 | NP_898897.1 | 1 - 2 | 304 | 92 | |
GenBank mRNA | AB082517.1 | - | 1 - 2 | 304 | 92 | |
BC162293.1 | - | 1 - 2 | 298 | 92 | ||
BC162308.1 | - | 1 - 2 | 298 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |