Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199909.2 | NP_956203.1 | 6 - 7 | 579 | 70 | |
GenBank mRNA | AY162326.1 | - | 6 - 7 | 579 | 70 | |
BC053124.1 | - | 6 - 7 | 560 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |