Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199909.2 | NP_956203.1 | 7 - 8 | 685 | 136 | |
GenBank mRNA | AY162326.1 | - | 7 - 8 | 685 | 136 | |
BC053124.1 | - | 7 - 8 | 666 | 136 | ||
BC150356.1 | - | 7 - 8 | 689 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |