Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199928.1 | NP_956222.1 | 6 - 7 | 667 | 92 | |
GenBank mRNA | BC044514.1 | - | 6 - 7 | 667 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |