Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crispld1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200470.1 | NP_956764.1 | 10 - 11 | 1403 | 74 | |
XM_009298567.3 | XP_009296842.1 | 14 - 15 | 1744 | 74 | ||
GenBank mRNA | BC055176.1 | - | 10 - 11 | 1403 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |