Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc15a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC049320.1 | - | 7 - 8 | 1295 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |