Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201124.1 | NP_957418.1 | 3 - 4 | 247 | 143 | |
XM_009303041.3 | XP_009301316.1 | 3 - 4 | 312 | 143 | ||
GenBank mRNA | BC054595.1 | - | 3 - 4 | 247 | 143 | |
BC164508.1 | - | 3 - 4 | 142 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |