Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201171.1 | NP_957465.1 | 2 - 3 | 648 | 111 | |
GenBank mRNA | BC052477.1 | - | 2 - 3 | 648 | 111 | |
BC164196.1 | - | 2 - 3 | 535 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |