Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plpp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205660.1 | NP_991223.1 | 1 - 2 | 547 | 69 | |
GenBank mRNA | BC065648.1 | - | 1 - 2 | 547 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |