Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grhprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205685.2 | NP_991248.2 | 6 - 7 | 901 | 139 | |
XM_005168340.4 | XP_005168397.1 | 6 - 7 | 866 | 139 | ||
GenBank mRNA | BC052108.1 | - | 6 - 7 | 710 | 139 | |
BC065431.1 | - | 6 - 7 | 761 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |