Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dipk1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212751.1 | NP_997916.1 | 3 - 4 | 1216 | 68 | |
GenBank mRNA | BC067726.1 | - | 3 - 4 | 1216 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |