Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kirrel1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001362419.1 | NP_001349348.1 | 11 - 12 | 1661 | 93 | |
NM_212868.2 | NP_998033.1 | 11 - 12 | 1661 | 93 | ||
GenBank mRNA | BC066766.1 | - | 11 - 12 | 1661 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |