Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: laptm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214685.2 | NP_999850.2 | 5 - 6 | 593 | 73 | |
GenBank mRNA | BC058878.1 | - | 5 - 6 | 620 | 73 | |
BC155748.1 | - | 5 - 6 | 593 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |