Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpm6ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214687.1 | NP_999852.1 | 2 - 3 | 232 | 65 | |
XM_005159981.4 | XP_005160038.1 | 2 - 3 | 725 | 65 | ||
XM_005159982.3 | XP_005160039.1 | 2 - 3 | 308 | 65 | ||
GenBank mRNA | AY070264.1 | - | 2 - 3 | 232 | 65 | |
AY070266.1 | - | 2 - 3 | 232 | 65 | ||
BC075774.1 | - | 2 - 3 | 316 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |