Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thrap3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214815.1 | NP_999980.1 | 7 - 8 | 2056 | 104 | |
XM_005173742.4 | XP_005173799.1 | 8 - 9 | 2035 | 104 | ||
XM_021468043.1 | XP_021323718.1 | 7 - 8 | 2168 | 104 | ||
XM_021468044.1 | XP_021323719.1 | 5 - 6 | 1096 | 104 | ||
XM_021468045.1 | XP_021323720.1 | 7 - 8 | 1344 | 104 | ||
GenBank mRNA | BC070004.1 | - | 7 - 8 | 2056 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |