Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: parp6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002537.1 | NP_001002537.1 | 3 - 4 | 353 | 74 | |
XM_021467633.1 | XP_021323308.1 | 3 - 4 | 489 | 74 | ||
GenBank mRNA | BC076271.1 | - | 3 - 4 | 353 | 74 |
Gene Symbol: LOC100537098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017351978.2 | XP_017207467.2 | 401 | 74 | ||
XM_021467719.1 | XP_021323394.1 | 401 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |